accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NHOOO

CAS RN 2568-34-5

IUPAC Name: 2-(N-methyl-1-phenylformamido)acetic acid
Synonyms: (N-methyl-1-phenylformamido)acetic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: PKCSYDDSNIJRIX-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)C(=O)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results