accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NaOSOONHNNSOOO

CAS RN 25641-18-3

IUPAC Name: sodium 10-phenyl-12-[(4-sulfonatophenyl)amino]-5,10λ⁵-diazatetraphen-10-ylium-11-sulfonate
Synonyms: sodium 10-phenyl-12-[(4-sulfonatophenyl)amino]-5,10λ⁵-diazatetraphen-10-ylium-11-sulfonate
Molecular Weight (g/mol): 579.58
Molecular Formula: C28H18N3NaO6S2
InChi Key: WORJHAQCTLEJBQ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(NC2=C(C3=C(N=C4C=CC=CC4=[N+]3C3=CC=CC=C3)C3=CC=CC=C23)S([O-])(=O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)