accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FH2NI

CAS RN 255724-71-1

IUPAC Name: 5-fluoro-2-iodoaniline
Synonyms: 5-fluoro-2-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: UCPDOOZBROQHME-UHFFFAOYSA-N
SMILES: NC1=C(I)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results