accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 25519-78-2

IUPAC Name: hydrogen 4-(4-fluorobenzoyl)piperidine chloride
Synonyms: hydrogen 4-(4-fluorobenzoyl)piperidine chloride
Molecular Weight (g/mol): 243.71
Molecular Formula: C12H15ClFNO
InChi Key: GPKDBZQZPNOBGM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results