accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OSiH2COH3COH3C

CAS RN 2551-83-9

IUPAC Name: trimethoxy(prop-2-en-1-yl)silane
Synonyms: trimethoxy(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 162.26
Molecular Formula: C6H14O3Si
InChi Key: LFRDHGNFBLIJIY-UHFFFAOYSA-N
SMILES: CO[Si](CC=C)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results