accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOOOCs

CAS RN 2550-61-0

IUPAC Name: caesium(1+) methanesulfonate
Synonyms: caesium(1+) mesylate
Molecular Weight (g/mol): 228
Molecular Formula: CH3CsO3S
InChi Key: DJKJXRLREATOMF-UHFFFAOYSA-M
SMILES: [Cs+].CS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results