accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOK

CAS RN 254966-93-3

IUPAC Name: potassium acetate hydrate
Synonyms: potassium acetate hydrate
Molecular Weight (g/mol): 116.16
Molecular Formula: C2H5KO3
InChi Key: YCYVPVNGUXMWMP-UHFFFAOYSA-M
SMILES: O.[K+].CC([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results