accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClNOHO

CAS RN 25462-85-5

IUPAC Name: 2-chloro-6-methylpyridine-4-carboxylic acid
Synonyms: 2-chloro-6-methylpyridine-4-carboxylic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: ZGZMEKHQIZSZOH-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(Cl)=N1)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)