accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNCl

CAS RN 253453-91-7

IUPAC Name: 2-chloro-1-methyl-1H-imidazole
Synonyms: 2-chloro-1-methylimidazole
Molecular Weight (g/mol): 116.55
Molecular Formula: C4H5ClN2
InChi Key: VSCQFRRKWFQPOA-UHFFFAOYSA-N
SMILES: CN1C=CN=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results