accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CBrOH2CBrBrOH2CBr

CAS RN 25327-89-3

IUPAC Name: 1,3-dibromo-5-{2-[3,5-dibromo-4-(prop-2-en-1-yloxy)phenyl]propan-2-yl}-2-(prop-2-en-1-yloxy)benzene
Synonyms: tetrabromobisphenol A bis(allyl ether)
Molecular Weight (g/mol): 624.01
Molecular Formula: C21H20Br4O2
InChi Key: PWXTUWQHMIFLKL-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC(Br)=C(OCC=C)C(Br)=C1)C1=CC(Br)=C(OCC=C)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results