accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOFOOCH3

CAS RN 252938-22-0

IUPAC Name: 1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Synonyms: 1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Molecular Weight (g/mol): 240.23
Molecular Formula: C12H13FO4
InChi Key: WDFUURBBGZTSIA-UHFFFAOYSA-N
SMILES: COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results