accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2528-00-9

IUPAC Name: ethyl 5-(chloromethyl)furan-2-carboxylate
Synonyms: ethyl 5-(chloromethyl)furan-2-carboxylate
Molecular Weight (g/mol): 188.61
Molecular Formula: C8H9ClO3
InChi Key: JBACYJRMCXLIQU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(CCl)O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results