accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CON

CAS RN 2525-16-8

IUPAC Name: 7-methoxy-3,4,5,6-tetrahydro-2H-azepine
Synonyms: methyl tetrahydroazepinyl ether
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: DNXIQMQGKSQHPC-UHFFFAOYSA-N
SMILES: COC1=NCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results