accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOHH2N

CAS RN 2521-84-8

IUPAC Name: 2-amino-2-cyclopentylacetic acid
Synonyms: cyclopentylglycine
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: XBPKRVHTESHFAA-UHFFFAOYNA-N
SMILES: NC(C1CCCC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)