accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 25186-43-0

CH3H3CNHNOO

CAS RN 25186-43-0

IUPAC Name: 4-nitro-N-(propan-2-yl)aniline
Synonyms: N-isopropyl-4-nitroaniline
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: VTSUWHFLMJLYKN-UHFFFAOYSA-N
SMILES: CC(C)NC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results