CAS RN 25173-72-2
IUPAC Name:
3-(3,4,5-trimethoxyphenyl)propanoic acid
Synonyms:
3-(3,4,5-trimethoxyphenyl)propanoic acid
Molecular Weight (g/mol):
240.26
Molecular Formula:
C12H16O5
InChi Key:
ZCYXGVJUZBKJAI-UHFFFAOYSA-N
SMILES:
COC1=CC(CCC(O)=O)=CC(OC)=C1OC
1
–
2
of
2
results