accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSOOOHO

CAS RN 2516-97-4

IUPAC Name: 2-methanesulfonylacetic acid
Synonyms: methanesulfonylacetic acid
Molecular Weight (g/mol): 138.14
Molecular Formula: C3H6O4S
InChi Key: NYEHUAQIJXERLP-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)