CAS RN 2514708-84-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-(1-Oxidobenzo[d]isothiazol-3-yl)piperazine 1-Oxide Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(1-Oxidobenzo[d]isothiazol-3-yl)piperazine 1-Oxide Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(1-Oxidobenzo[d]isothiazol-3-yl)piperazine 1-Oxide Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.