CAS RN 2512215-78-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Thiazol-5-ylmethyl ((2S,3S,5S)-5-Amino-3-((tert-butyldimethylsilyl)oxy)-1,6-diphenylhexan-2-yl)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Thiazol-5-ylmethyl ((2S,3S,5S)-5-Amino-3-((tert-butyldimethylsilyl)oxy)-1,6-diphenylhexan-2-yl)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Thiazol-5-ylmethyl ((2S,3S,5S)-5-Amino-3-((tert-butyldimethylsilyl)oxy)-1,6-diphenylhexan-2-yl)carbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.