CAS RN 2512209-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-tert-Butyl 3-Ethyl 5-methyl 2-((2-((tert-Butoxycarbonyl)amino)ethoxy)methyl)-4-(2-chlorophenyl)-6-methylpyridine-1,3,5(4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-tert-Butyl 3-Ethyl 5-methyl 2-((2-((tert-Butoxycarbonyl)amino)ethoxy)methyl)-4-(2-chlorophenyl)-6-methylpyridine-1,3,5(4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-tert-Butyl 3-Ethyl 5-methyl 2-((2-((tert-Butoxycarbonyl)amino)ethoxy)methyl)-4-(2-chlorophenyl)-6-methylpyridine-1,3,5(4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.