CAS RN 2512202-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-((tert-Butyldimethylsilyl)oxy)-2,5-bis(trifluoromethyl)aniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-((tert-Butyldimethylsilyl)oxy)-2,5-bis(trifluoromethyl)aniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.