CAS RN 2510-71-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-9,10-Dihydroxy-9,10-dihydrophenanthrene, TRC
CAS: 2510-71-6 Molecular Formula: C14H12O2 Molecular Weight (g/mol): 212.24 Synonym: (9R,10S)-rel-9,10-Dihydro-9,10-dihydroxyphenanthrene,9,10-Dihydrophenanthrene-cis-9,10-diol,Phenanthrene cis-9,10-Hihydrodiol,cis-9,10-Dihydro-9,10-phenanthrenediol SMILES: O[C@@H]1[C@H](O)c2ccccc2c3ccccc13