CAS RN 25086-89-9
IUPAC Name:
1-ethenylpyrrolidin-2-one; ethenyl acetate
Synonyms:
VP/VA copolymer
Molecular Weight (g/mol):
197.23
Molecular Formula:
C10H15NO3
InChi Key:
FYUWIEKAVLOHSE-UHFFFAOYSA-N
SMILES:
CC(=O)OC=C.C=CN1CCCC1=O
1
–
3
of
3
results
1
Copovidone (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Copovidone (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Copovidone (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.