CAS RN 25041-66-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Aurantinidin Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.