accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2500-88-1

IUPAC Name: 1-(octadecyldisulfanyl)octadecane
Synonyms: 1-(octadecyldisulfanyl)octadecane
Molecular Weight (g/mol): 571.11
Molecular Formula: C36H74S2
InChi Key: MQQKTNDBASEZSD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCSSCCCCCCCCCCCCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results