accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2491-32-9

CAS RN 2491-32-9

IUPAC Name: 1-(4-hydroxyphenyl)-2-phenylethan-1-one
Synonyms: 1-(4-hydroxyphenyl)-2-phenylethanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: JBQTZLNCDIFCCO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results