accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 24447-99-2

H3CNHSOHOO

CAS RN 24447-99-2

IUPAC Name: 4-(methylamino)benzene-1-sulfonic acid
Synonyms: 4-(methylamino)benzenesulfonic acid
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: QRAXZXPSAGQUNP-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results