accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2443-66-5

IUPAC Name: 2-hydroxy-2-phenylacetohydrazide
Synonyms: 2-hydroxy-2-phenylacetohydrazide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: FWTGUGVETHVGTL-UHFFFAOYNA-N
SMILES: NNC(=O)C(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results