accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOSFFF

CAS RN 243977-23-3

IUPAC Name: 2-{4-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Synonyms: {4-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Molecular Weight (g/mol): 236.21
Molecular Formula: C9H7F3O2S
InChi Key: BFDKCZMYQOSTJG-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(SC(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results