accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 24393-53-1

H3COOBr(E)

CAS RN 24393-53-1

IUPAC Name: ethyl (2E)-3-(4-bromophenyl)prop-2-enoate
Synonyms: ethyl (2E)-3-(4-bromophenyl)prop-2-enoate
Molecular Weight (g/mol): 255.11
Molecular Formula: C11H11BrO2
InChi Key: YOOKYIPLSLPRTC-VMPITWQZSA-N
SMILES: CCOC(=O)\C=C\C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results