accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNHCH3

CAS RN 2439-54-5

IUPAC Name: methyl(octyl)amine
Synonyms: methyl(octyl)amine
Molecular Weight (g/mol): 143.27
Molecular Formula: C9H21N
InChi Key: SEGJNMCIMOLEDM-UHFFFAOYSA-N
SMILES: CCCCCCCCNC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)