accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 24386-17-2

NH2OOON

CAS RN 24386-17-2

IUPAC Name: 5'-amino-2'-(furan-2-yl)-[2,3'-bifuran]-4'-carbonitrile
Synonyms: 5'-amino-2'-(furan-2-yl)-[2,3'-bifuran]-4'-carbonitrile
Molecular Weight (g/mol): 240.22
Molecular Formula: C13H8N2O3
InChi Key: RDQSOEAGWZLHBE-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C(C2=CC=CO2)=C(O1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results