accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 24134-12-1

IUPAC Name: 1,2-dimethyl-1H-imidazole-5-carbaldehyde
Synonyms: 2,3-dimethylimidazole-4-carbaldehyde
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: XEMFQOKIINBGDI-UHFFFAOYSA-N
SMILES: CN1C(C)=NC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results