accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 239135-49-0

NH2FFFF

CAS RN 239135-49-0

IUPAC Name: 1-[2-fluoro-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-fluoro-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: PQUYJDPJHOSOTN-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results