accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 239074-29-4

H3CH3CCH3OONHOH

CAS RN 239074-29-4

IUPAC Name: tert-butyl N-[4-(hydroxymethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(hydroxymethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 229.32
Molecular Formula: C12H23NO3
InChi Key: SGNKPJPMWHKOJO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CO)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)