accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOCH3(E)

CAS RN 2390-09-2

IUPAC Name: methyl (11E)-icos-11-enoate
Synonyms: methyl (11E)-icos-11-enoate
Molecular Weight (g/mol): 324.55
Molecular Formula: C21H40O2
InChi Key: RBKMRGOHCLRTLZ-ZHACJKMWSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)