accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23897-15-6

CH3H3CPH3CH3CCH3H3CCH3CH3CH3

CAS RN 23897-15-6

IUPAC Name: tris(2,4,6-trimethylphenyl)phosphane
Synonyms: tris(2,4,6-trimethylphenyl)phosphane
Molecular Weight (g/mol): 388.54
Molecular Formula: C27H33P
InChi Key: IDXDWPWXHTXJMZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(P(C2=C(C)C=C(C)C=C2C)C2=C(C)C=C(C)C=C2C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results