accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2389-48-2

IUPAC Name: methyl 2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Synonyms: methyl 2-amino-6-[(tert-butoxycarbonyl)amino]hexanoate hydrochloride
Molecular Weight (g/mol): 296.79
Molecular Formula: C12H25ClN2O4
InChi Key: NANRHOPPXCBHGI-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CCCCNC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)