accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 238749-50-3

IUPAC Name: ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate
Synonyms: ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate
Molecular Weight (g/mol): 274.13
Molecular Formula: C9H8BrNO2S
InChi Key: KXGSKTSSCQBDOA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=C(N1)C=C(Br)S2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results