accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONH2N

CAS RN 23842-82-2

IUPAC Name: 2-amino-5-methoxybenzonitrile
Synonyms: 2-amino-5-methoxybenzonitrile
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: SRWMPAZUWXLIPG-UHFFFAOYSA-N
SMILES: COC1=CC(C#N)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results