accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOCl

CAS RN 23808-46-0

IUPAC Name: 5-(2-chloroethyl)-2H-1,3-benzodioxole
Synonyms: 5-(2-chloroethyl)-2H-1,3-benzodioxole
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: YFVIYUCTLBXCMJ-UHFFFAOYSA-N
SMILES: ClCCC1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results