accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23761-23-1

OOHO

CAS RN 23761-23-1

IUPAC Name: 3-oxocyclobutane-1-carboxylic acid
Synonyms: 3-oxocyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: IENOFRJPUPTEMI-UHFFFAOYSA-N
SMILES: OC(=O)C1CC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results