accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 237385-98-7

CH3OOSFFF

CAS RN 237385-98-7

IUPAC Name: methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Synonyms: methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 286.27
Molecular Formula: C13H9F3O2S
InChi Key: RDMSDMSSVFMCKQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(S1)C(F)(F)F)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results