accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SCl

CAS RN 2373-51-5

IUPAC Name: chloro(methylsulfanyl)methane
Synonyms: chloromethyl methyl sulfide
Molecular Weight (g/mol): 96.57
Molecular Formula: C2H5ClS
InChi Key: JWMLCCRPDOIBAV-UHFFFAOYSA-N
SMILES: CSCCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results