accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFFFFFF

CAS RN 237069-82-8

IUPAC Name: 1-[2,4-bis(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[2,4-bis(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 256.15
Molecular Formula: C10H6F6O
InChi Key: MHLYNSLYUWMONA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results