accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2FNOO

CAS RN 2369-11-1

IUPAC Name: 5-fluoro-2-nitroaniline
Synonyms: 5-fluoro-2-nitroaniline
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: PEDMFCHWOVJDNW-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results