accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2365-85-7

NH2FOHO

CAS RN 2365-85-7

IUPAC Name: 3-amino-4-fluorobenzoic acid
Synonyms: 3-amino-4-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: WFSPEVFSRUTRCN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results