CAS RN 2358-29-4
CAS RN 2358-29-4
IUPAC Name:
2,3,6-trifluorobenzoate
Synonyms:
2,3,6-trifluorobenzoate
Molecular Weight (g/mol):
175.09
Molecular Formula:
C7H2F3O2
InChi Key:
MGUPHQGQNHDGNK-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=C(F)C=CC(F)=C1F
1
–
1
of
1
results
1
2,3,6-Trifluorobenzoic acid, 99%, Thermo Scientific™
CAS: 2358-29-4 Molecular Formula: C7H2F3O2 Molecular Weight (g/mol): 175.09 MDL Number: MFCD00002409 InChI Key: MGUPHQGQNHDGNK-UHFFFAOYSA-M Synonym: 2,5,6-trifluorobenzoic acid,2,3,6-trifluorobenzoicacid,2,3,6-trifluoro-benzoic acid,benzoic acid, 2,3,6-trifluoro,pubchem1344,intermediates-zcf02628,ksc494k2n,2,3,6-trifluoro benzoic acid,rarechem al bo 0267,2,3,6-trifluorobenzoic acid PubChem CID: 520062 SMILES: [O-]C(=O)C1=C(F)C=CC(F)=C1F