accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 235088-69-4

H2NFFF

CAS RN 235088-69-4

IUPAC Name: 1-(3,4,5-trifluorophenyl)methanamine
Synonyms: 1-(3,4,5-trifluorophenyl)methanamine
Molecular Weight (g/mol): 161.13
Molecular Formula: C7H6F3N
InChi Key: AJGBQAAXUVSBCH-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=C(F)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results