CAS RN 23496-41-5
CAS RN 23496-41-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Peimine, MedChemExpress
MedChemExpress Peimine(Dihydroisoimperialine; Verticine) is a natural compound with good anti-inflammatory effects in vivo.